This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other IFI44 SIRNA Assays ›
Gene Symbol:
IFI44
Gene Name:
interferon induced protein 44
Gene Aliases:
MTAP44, TLDC5, p44
Chromosome Location:
Chr.1: 78648554 - 78664078 on Build GRCh38
UniGene ID:
Hs.82316
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s20721

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: IFI44
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006417.4 NP_006408.3 5 948
NR_135640.1 4 484
NR_135641.1 5 951
XM_005270380.3 XP_005270437.1 5 2186
XM_011540516.2 XP_011538818.1 5 2186
XM_017000083.1 XP_016855572.1 5 2186
GenBank mRNA AK302527.1 - 5 909
AK316278.1 - 4 441
AK316476.1 - 5 906
BC022870.1 - 5 913 4071404

Back To Top

More Information


Gene Ontology Categories:

Function(s)

response to virus

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-ml9gc:80/100.66.72.222:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0