siRNA Details
| Gene Symbol: FAM189B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267608.1 | NP_001254537.1 | 9 | 2078 | ||
| NM_006589.2 | NP_006580.2 | 10 | 2132 | |||
| NM_198264.1 | NP_937995.1 | 7 | 1844 | |||
| XM_005244845.1 | XP_005244902.1 | 10 | 2129 | |||
| GenBank mRNA | AF070550.1 | - | 4 | 527 | ||
| AL353941.1 | - | 5 | 916 | |||
| BC006297.1 | - | 2 | 544 | |||
| BC006493.2 | - | 10 | 1612 | 2966515 | ||
| BC007444.2 | - | 10 | 1612 | 2966515 | ||
| BC010471.1 | - | 10 | 2036 | 4179576 | ||
| BC017262.2 | - | 3 | 663 | |||
| BC040136.1 | - | 7 | 1321 | 6061151 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001014995, BC068123, BC085505, BC088990.

FAM189B
Genomic Map