siRNA Details
| Gene Symbol: Aloxe3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011786.2 | NP_035916.2 | 1 | 84 | ||
| GenBank mRNA | AK141003.1 | - | 1 | 247 | ||
| BC116258.1 | - | 1 | 83 | 40093733 | ||
| BC116259.1 | - | 1 | 83 | 40093735 | ||
| Y14695.1 | - | 1 | 84 | |||

Aloxe3
Genomic Map