This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other BLCAP SIRNA Assays ›
Gene Symbol:
BLCAP
Gene Name:
bladder cancer associated protein
Gene Aliases:
BC10
Chromosome Location:
Chr.20: 37517417 - 37527931 on Build GRCh38
UniGene ID:
Hs.472651
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s21431

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: BLCAP
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167820.1 NP_001161292.1 3 1556
NM_001167821.1 NP_001161293.1 2 1386
NM_001167822.2 NP_001161294.1 2 1443
NM_001167823.1 NP_001161295.1 2 1459
NM_001317074.1 NP_001304003.1 2 1436
NM_001317075.1 NP_001304004.1 2 1352
NM_006698.3 NP_006689.1 2 1423
XM_017027599.1 XP_016883088.1 2 2336
GenBank mRNA AF053470.1 - 2 1362
AK074498.1 - 5 1172
AK124822.1 - 3 1281
AL049288.1 - 2 1369
BC047692.1 - 2 1354 6059195
BC071704.1 - 2 1348 5190373

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell cycle
apoptotic nuclear changes
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0