This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TLK2 SIRNA Assays ›
Gene Symbol:
TLK2
Gene Name:
tousled like kinase 2
Gene Aliases:
HsHPK, PKU-ALPHA
Chromosome Location:
Chr.17: 62477481 - 62615481 on Build GRCh38
UniGene ID:
Hs.445078
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s21678
Validated

Purification
Size
Availability Inventoried
Catalog # 4390824
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TLK2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284333.1 NP_001271262.1 18, 19 1888
NM_001284363.1 NP_001271292.1 15, 16 1483
NM_006852.3 NP_006843.2 17, 18 1825
XM_011524215.2 XP_011522517.1 16, 17 1532
XM_011524216.2 XP_011522518.1 18, 19 1652
XM_011524217.2 XP_011522519.1 18, 19 1839
XM_011524218.2 XP_011522520.1 15, 16 1466
XM_011524220.2 XP_011522522.1 15, 16 1436
XM_011524221.2 XP_011522523.1 17, 18 1550
XM_011524222.2 XP_011522524.1 18, 19 2031
XM_011524223.2 XP_011522525.1 16, 17 1693
XM_011524224.2 XP_011522526.1 16, 17 1751
XM_011524225.2 XP_011522527.1 16, 17 1699
XM_011524226.2 XP_011522528.1 18, 19 1745
XM_011524227.2 XP_011522529.1 17, 18 1799
XM_011524228.2 XP_011522530.1 18, 19 1899
XM_011524229.2 XP_011522531.1 19, 20 1826
XM_017024044.1 XP_016879533.1 17, 18 1659
XM_017024045.1 XP_016879534.1 16, 17 1767
XM_017024046.1 XP_016879535.1 17, 18 1589
XM_017024047.1 XP_016879536.1 17, 18 1586
XM_017024048.1 XP_016879537.1 17, 18 2047
XM_017024049.1 XP_016879538.1 17, 18 2000
XM_017024050.1 XP_016879539.1 15, 16 1532
XM_017024051.1 XP_016879540.1 16, 17 1981
XM_017024052.1 XP_016879541.1 16, 17 1932
XM_017024053.1 XP_016879542.1 16, 17 1604
XM_017024054.1 XP_016879543.1 15, 16 1630
XM_017024055.1 XP_016879544.1 14, 15 1534
XM_017024056.1 XP_016879545.1 15, 16 1685
XM_017024057.1 XP_016879546.1 15, 16 1499
XM_017024058.1 XP_016879547.1 17, 18 1777
XM_017024059.1 XP_016879548.1 17, 18 1767
XM_017024060.1 XP_016879549.1 16, 17 1684
GenBank mRNA AB004884.1 - 15, 16 1461
AF162667.1 - 17, 18 1697
AK300586.1 - 15, 16 1483
AX775745.1 - 17, 18 1697
BC044925.1 - 18, 19 1795

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001191652, XM_006247576, XM_008768350, XM_006247574, XM_008768348, and others.
This siRNA also targets the following transcripts in Mouse: NM_001294334, NM_001112705, NM_001294330, NM_001294331, NM_001294329, and others.

Matching Taqman Gene Expression Assay:


Hs00851784_g1

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

regulation of chromatin assembly or disassembly
protein phosphorylation
cellular response to DNA damage stimulus
cell cycle
chromosome segregation
negative regulation of autophagy
covalent chromatin modification
peptidyl-serine phosphorylation
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
intracellular signal transduction
cellular response to gamma radiation
protein serine/threonine kinase activity
protein binding
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-kbrl8:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline