This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RPS14 SIRNA Assays ›
Gene Symbol:
Rps14 LOC100911847
Gene Name:
ribosomal protein S14
40S ribosomal protein S14-like
Gene Aliases:
-
-
Chromosome Location:
Chr.18: 56042532 - 56047316 on Build Rnor_6.0
Chromosome Location:
Chr.5: 144083043 - 144083596 on Build Rnor_6.0
UniGene ID:
Rn.154712
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s218068

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rps14
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022672.1 NP_073163.1 3 318
GenBank mRNA BC058472.1 - 2 325 6918325
FQ210177.1 - 3 320
FQ210421.1 - 3 320
FQ210469.1 - 3 317
FQ211405.1 - 3 318
FQ211449.1 - 3 318
FQ211623.1 - 3 320
FQ212082.1 - 3 322
FQ212093.1 - 3 318
FQ212392.1 - 3 318
FQ214636.1 - 3 320
FQ217229.1 - 3 318
FQ217605.1 - 3 318
FQ218082.1 - 3 315
FQ218760.1 - 3 320
FQ219972.1 - 3 318
FQ220057.1 - 3 318
FQ220332.1 - 3 320
FQ220676.1 - 3 322
FQ220975.1 - 3 315
FQ221057.1 - 3 318
FQ221058.1 - 3 318
FQ221066.1 - 3 318
FQ221125.1 - 3 320
FQ221205.1 - 3 318
FQ221325.1 - 3 318
FQ221335.1 - 3 318
FQ221375.1 - 3 318
FQ221450.1 - 3 318
FQ221468.1 - 3 318
FQ221581.1 - 3 317
FQ221634.1 - 3 315
FQ221754.1 - 3 314
FQ221769.1 - 3 320
FQ221835.1 - 3 320
FQ221893.1 - 3 320
FQ221920.1 - 3 320
FQ222066.1 - 3 315
FQ222119.1 - 3 318
FQ222124.1 - 3 318
FQ222159.1 - 3 320
FQ222163.1 - 3 319
FQ222324.1 - 3 322
FQ222370.1 - 3 318
FQ222480.1 - 3 318
FQ222552.1 - 3 317
FQ222574.1 - 3 318
FQ222575.1 - 2 318
FQ222601.1 - 3 332
FQ222667.1 - 3 318
FQ222685.1 - 3 318
FQ222697.1 - 3 320
FQ222714.1 - 3 318
FQ222747.1 - 3 322
FQ222803.1 - 3 318
FQ222840.1 - 3 321
FQ222893.1 - 3 320
FQ222979.1 - 3 320
FQ222986.1 - 3 315
FQ223007.1 - 3 318
FQ223012.1 - 3 320
FQ223069.1 - 3 320
FQ223131.1 - 3 318
FQ223204.1 - 3 320
FQ223283.1 - 3 318
FQ223454.1 - 3 319
FQ223468.1 - 3 318
FQ223480.1 - 3 318
FQ223519.1 - 3 317
FQ223527.1 - 3 318
FQ223570.1 - 3 318
FQ224051.1 - 3 315
FQ224105.1 - 3 320
FQ224302.1 - 3 319
FQ224493.1 - 3 320
FQ224808.1 - 3 322
FQ225464.1 - 3 320
FQ228360.1 - 3 303
FQ228480.1 - 3 320
FQ228486.1 - 3 318
FQ228559.1 - 3 322
FQ228608.1 - 3 318
FQ228692.1 - 3 317
FQ229006.1 - 3 324
FQ229014.1 - 3 320
FQ229055.1 - 3 320
FQ229061.1 - 3 320
FQ229229.1 - 3 320
FQ229387.1 - 3 318
FQ231306.1 - 461
X15040.1 - 3 318
Gene Symbol: LOC100911847
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750030.3 XP_003750078.3 1 322

Back To Top

More Information


Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

ribosomal small subunit assembly
negative regulation of transcription from RNA polymerase II promoter
translation
regulation of translation
erythrocyte differentiation
maturation of SSU-rRNA
RNA binding
structural constituent of ribosome
poly(A) RNA binding
translation regulator activity
mRNA 5'-UTR binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-bthjr:80/100.66.76.203:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline