This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FAM107A SIRNA Assays ›
Gene Symbol:
FAM107A
Gene Name:
family with sequence similarity 107 member A
Gene Aliases:
DRR1, TU3A
Chromosome Location:
Chr.3: 58564112 - 58627610 on Build GRCh38
UniGene ID:
Hs.506357
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s22042

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 388.00
Your Price
Online offer: 287.12
287.12
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM107A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001076778.2 NP_001070246.1 4 1608
NM_001282713.1 NP_001269642.1 4 1408
NM_001282714.1 NP_001269643.1 5 1856
NM_007177.3 NP_009108.1 5 1706
GenBank mRNA AB023811.1 - 4 1186
AF035283.1 - 4 1607
AF089853.1 - 4 1697
AF089854.1 - 5 1795
AK054720.1 - 5 1856
AK055443.1 - 3 1744
AK128791.1 - 4 1235
AK298001.1 - 4 1363
AK316450.1 - 4 1263
AL050264.1 - 4 1221
BC010561.1 - 5 1307 4152118
EU569833.1 - 4 1263

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline