This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PDS5A SIRNA Assays ›
Gene Symbol:
Pds5a
Gene Name:
PDS5 cohesin associated factor A
Gene Aliases:
RGD1307094, Scc-112
Chromosome Location:
Chr.14: 44185445 - 44282308 on Build Rnor_6.0
UniGene ID:
Rn.224580
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s221025

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pds5a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083624.1 NP_001077093.1 6 712
XM_008770153.1 XP_008768375.1 6 746
XM_008770154.1 XP_008768376.1 5 659
GenBank mRNA EF460313.1 - 6 712

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001081321, AK172992.

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s)

mitotic sister chromatid cohesion
negative regulation of DNA replication
cell division

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-hssmq:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline