siRNA Details
| Gene Symbol: Derl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014202.1 | NP_001014224.1 | 5 | 469 | ||
| GenBank mRNA | BC085877.1 | - | 5 | 469 | 7191715 | |
| FQ218639.1 | - | 5 | 506 | |||
| FQ220628.1 | - | 5 | 508 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Derl1| Gene Symbol: Derl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014202.1 | NP_001014224.1 | 5 | 469 | ||
| GenBank mRNA | BC085877.1 | - | 5 | 469 | 7191715 | |
| FQ218639.1 | - | 5 | 506 | |||
| FQ220628.1 | - | 5 | 508 | |||