siRNA Details
| Gene Symbol: Hormad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108949.1 | NP_001102419.1 | 11, 12 | 949 | ||
| XM_006232951.2 | XP_006233013.1 | 10, 11 | 904 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hormad1| Gene Symbol: Hormad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108949.1 | NP_001102419.1 | 11, 12 | 949 | ||
| XM_006232951.2 | XP_006233013.1 | 10, 11 | 904 | |||