siRNA Details
| Gene Symbol: Mrgprb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_577804.4 | XP_577804.2 | 1 | 624 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrgprb2| Gene Symbol: Mrgprb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_577804.4 | XP_577804.2 | 1 | 624 | ||
Set Membership: |
|