siRNA Details
| Gene Symbol: Abracl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099647.1 | NP_001093117.1 | 3 | 351 | ||
| GenBank mRNA | BC089921.1 | - | 1 | 584 | 7306744 | |
| CO569536.1 | - | 3 | 351 | |||
| FQ221434.1 | - | 3 | 414 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abracl| Gene Symbol: Abracl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099647.1 | NP_001093117.1 | 3 | 351 | ||
| GenBank mRNA | BC089921.1 | - | 1 | 584 | 7306744 | |
| CO569536.1 | - | 3 | 351 | |||
| FQ221434.1 | - | 3 | 414 | |||