siRNA Details
| Gene Symbol: Fam150b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109372.1 | NP_001102842.1 | 5 | 1306 | ||
| XM_006239971.2 | XP_006240033.1 | 4 | 1446 | |||
| GenBank mRNA | BC167018.1 | - | 5 | 1347 | 7307720 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam150b| Gene Symbol: Fam150b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109372.1 | NP_001102842.1 | 5 | 1306 | ||
| XM_006239971.2 | XP_006240033.1 | 4 | 1446 | |||
| GenBank mRNA | BC167018.1 | - | 5 | 1347 | 7307720 | |