This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FOXK2 SIRNA Assays ›
Gene Symbol:
Foxk2
Gene Name:
forkhead box K2
Gene Aliases:
-
Chromosome Location:
Chr.10: 110468893 - 110518854 on Build Rnor_6.0
UniGene ID:
Rn.44253
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s222632

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Foxk2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006247890.2 XP_006247952.1 8 1766
GenBank mRNA BC168919.1 - 7 1234

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001080932, EU882160.

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
magnesium ion binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-mjcrx:80/100.66.74.196:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0