siRNA Details
| Gene Symbol: CYP4F8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007253.3 | NP_009184.1 | 12 | 1420 | ||
| XM_017026232.1 | XP_016881721.1 | 10 | 1292 | |||
| GenBank mRNA | AF133298.1 | - | 11 | 1363 | ||
| AK092612.1 | - | 1 | 1632 | |||
| AK096809.1 | - | 8 | 3074 | |||
| AK300530.1 | - | 10 | 1446 | |||
| AK300539.1 | - | 8 | 971 | |||
| AK314685.1 | - | 12 | 1418 | |||

CYP4F8
Genomic Map