This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PTMS SIRNA Assays ›
Gene Symbol:
Ptms
Gene Name:
parathymosin
Gene Aliases:
ZnBP
Chromosome Location:
Chr.4: 157435371 - 157439507 on Build Rnor_6.0
UniGene ID:
Rn.3609
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s222869

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ptms
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031975.2 NP_114181.1 4, 5 597
GenBank mRNA BC167753.1 - 4, 5 587 7371571
M20616.1 - 3, 4 201
M33025.1 - 3, 4 201
X16481.1 - 4, 5 380

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_002824, M24398, BC128229, BC017025, BM550529, and others.

Gene Ontology Categories:

Function(s) Process(es)

immune system process
negative regulation of catalytic activity
enzyme inhibitor activity
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0