This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other KCTD21 SIRNA Assays ›
Gene Symbol:
KCTD21
Gene Name:
potassium channel tetramerization domain containing 21
Gene Aliases:
KCASH2
Chromosome Location:
Chr.11: 78171243 - 78188854 on Build GRCh38
UniGene ID:
Hs.528439
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s223773

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KCTD21
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029859.2 NP_001025030.1 2 773
XM_005273925.3 XP_005273982.1 2 998
XM_005273928.1 XP_005273985.1 3 898
XM_006718516.3 XP_006718579.1 2 3943
XM_006718517.2 XP_006718580.1 3 1010
XM_006718518.3 XP_006718581.1 2 758
XM_011544955.2 XP_011543257.1 2 2238
GenBank mRNA AK095233.1 - 3 522
AK300320.1 - 2 760
BC036058.1 - 2 762 5277643

Back To Top

More Information


Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s)

protein homooligomerization

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline