siRNA Details
| Gene Symbol: HPCAL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282396.1 | NP_001269325.1 | 5 | 1713 | ||
| NM_001282397.1 | NP_001269326.1 | 3 | 1391 | |||
| NM_016257.3 | NP_057341.1 | 4 | 1832 | |||
| XM_017001447.1 | XP_016856936.1 | 4 | 1445 | |||
| GenBank mRNA | AL136591.1 | - | 4 | 1539 | ||
| BC030827.1 | - | 4 | 1537 | 4811834 | ||

HPCAL4
Genomic Map