This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MAP3K19 SIRNA Assays ›
Gene Symbol:
MAP3K19
Gene Name:
mitogen-activated protein kinase kinase kinase 19
Gene Aliases:
RCK, YSK4
Chromosome Location:
Chr.2: 134963875 - 135047468 on Build GRCh38
UniGene ID:
Hs.659395
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s224847

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MAP3K19
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018046.2 NP_001018056.1 5 543
NM_001018047.2 NP_001018057.1 5 543
NM_001282883.1 NP_001269812.1 5 543
NM_001321177.1 NP_001308106.1 5 683
NM_025052.4 NP_079328.3 5 543
XM_011511891.2 XP_011510193.1 5 683
XM_011511892.1 XP_011510194.1 8 1251
XM_011511894.2 XP_011510196.1 5 658
XM_011511896.2 XP_011510198.1 5 524
XM_011511897.2 XP_011510199.1 5 604
XM_017005003.1 XP_016860492.1 8 1218
XM_017005004.1 XP_016860493.1 7 1103
XM_017005005.1 XP_016860494.1 6 904
XM_017005006.1 XP_016860495.1 5 683
XM_017005007.1 XP_016860496.1 5 683
GenBank mRNA AK122935.1 - 5 683
AK309454.1 - 7 1103
AY574901.1 - 5 543
AY574903.1 - 5 543
AY574904.1 - 5 543
AY574905.1 - 5 543
BC137276.1 - 5 542 9021273
BC137277.1 - 5 542 9021274
BC144555.1 - 5 542

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

signal transduction by protein phosphorylation
actin cytoskeleton organization
receptor signaling protein serine/threonine kinase activity
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0