This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CDSN SIRNA Assays ›
Gene Symbol:
CDSN
Gene Name:
corneodesmosin
Gene Aliases:
HTSS, HTSS1, HYPT2, PSS, PSS1, S
Chromosome Location:
Chr.6: 31115088 - 31120475 on Build GRCh38
UniGene ID:
Hs.310958
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s225430

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 388.00
Your Price
Online offer: 287.12
287.12
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CDSN
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001264.4 NP_001255.3 2 1002
GenBank mRNA AF030130.1 - 2 960
AK292636.1 - 2 970
BC031993.1 - 2 950 4747362
L20815.1 - 2 960

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
epidermis development
single organismal cell-cell adhesion
keratinocyte differentiation
skin morphogenesis
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline