This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CASC4 SIRNA Assays ›
Gene Symbol:
CASC4
Gene Name:
cancer susceptibility candidate 4
Gene Aliases:
H63
Chromosome Location:
Chr.15: 44288616 - 44415761 on Build GRCh38
UniGene ID:
Hs.512867
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s225491

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CASC4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138423.3 NP_612432.2 1, 2 645
NM_177974.2 NP_816929.1 1, 2 645
XM_017021880.1 XP_016877369.1 1, 2 740
GenBank mRNA AK291026.1 - 1, 2 402
AK298564.1 - 2, 3 360
AL832333.1 - 1, 2 417
AL832840.1 - 1, 2 87
AY358086.1 - 1, 2 596
BC012124.1 - 1, 2 625
BC063480.1 - 1, 2 599 5218924

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0