siRNA Details
| Gene Symbol: ZNRF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181710.3 | NP_859061.3 | 1 | 1160 | ||
| GenBank mRNA | AK098722.1 | - | 2 | 1027 | ||
| AK292251.1 | - | 1 | 1144 | |||
| BC017592.2 | - | 1 | 1144 | 4828408 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNRF4| Gene Symbol: ZNRF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181710.3 | NP_859061.3 | 1 | 1160 | ||
| GenBank mRNA | AK098722.1 | - | 2 | 1027 | ||
| AK292251.1 | - | 1 | 1144 | |||
| BC017592.2 | - | 1 | 1144 | 4828408 | ||