This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other WDR37 SIRNA Assays ›
Gene Symbol:
WDR37
Gene Name:
WD repeat domain 37
Gene Aliases:
-
Chromosome Location:
Chr.10: 1056836 - 1132297 on Build GRCh38
UniGene ID:
Hs.188495
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s22586

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: WDR37
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014023.3 NP_054742.2 3 358
GenBank mRNA AB023199.1 - 3 329
AK128194.1 - 2 3167
AK292591.1 - 3 321
AL136827.1 - 3 336
BC018044.1 - 3 387 4794184

Back To Top

More Information


Gene Ontology Categories:

Function(s)

maturation of 5.8S rRNA
maturation of LSU-rRNA

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline