This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMEM50A SIRNA Assays ›
Gene Symbol:
TMEM50A
Gene Name:
transmembrane protein 50A
Gene Aliases:
IFNRC, SMP1
Chromosome Location:
Chr.1: 25338298 - 25362361 on Build GRCh38
UniGene ID:
Hs.705699
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s225970

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM50A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014313.3 NP_055128.1 4 417
XM_005245817.1 XP_005245874.1 4 381
XM_011541159.1 XP_011539461.1 4 611
GenBank mRNA AF081282.1 - 4 344
AF087880.1 - 4 405
AK299180.1 - 4 381
AK311092.1 - 2 188
AK311355.1 - 2 162
AK312588.1 - 4 381
AL136627.1 - 4 399
AY071927.1 - 4 394
AY358650.1 - 4 380
BC007341.2 - 4 365 3688672
BC066908.1 - 4 356 4817784
JX644905.1 - 4 417
JX644906.1 - 4 417

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mlg7h:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline