This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other KIAA0825 SIRNA Assays ›
Gene Symbol:
KIAA0825
Gene Name:
KIAA0825
Gene Aliases:
C5orf36
Chromosome Location:
Chr.5: 94150851 - 94618604 on Build GRCh38
UniGene ID:
Hs.425123
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s226217

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KIAA0825
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145678.1 NP_001139150.1 5 901
NM_173665.2 NP_775936.1 5 901
XM_006714588.2 XP_006714651.1 5 901
XM_011543324.2 XP_011541626.1 5 901
XM_011543325.2 XP_011541627.1 6 967
XM_011543327.2 XP_011541629.1 5 817
XM_011543328.2 XP_011541630.1 6 1022
XM_011543329.2 XP_011541631.1 4 737
XM_011543330.2 XP_011541632.1 5 901
XM_011543331.1 XP_011541633.1 5 901
XM_011543332.1 XP_011541634.1 5 901
XM_011543333.2 XP_011541635.1 5 901
XM_011543334.2 XP_011541636.1 5 901
XM_011543335.2 XP_011541637.1 2 434
XM_017009373.1 XP_016864862.1 5 901
XM_017009374.1 XP_016864863.1 5 901
XM_017009375.1 XP_016864864.1 5 849
XM_017009376.1 XP_016864865.1 6 1017
XM_017009377.1 XP_016864866.1 4 743
GenBank mRNA AK308195.1 - 5 900
BC035515.1 - 5 875 5201671
BX648338.1 - 5 861

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline