siRNA Details
| Gene Symbol: FAM217B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190826.1 | NP_001177755.1 | 5 | 708 | ||
| NM_022106.2 | NP_071389.1 | 4 | 642 | |||
| XM_011528985.2 | XP_011527287.1 | 4 | 632 | |||
| XM_011528986.2 | XP_011527288.1 | 5 | 719 | |||
| GenBank mRNA | AK125073.1 | - | 5 | 708 | ||
| AL137442.1 | - | 1 | 92 | |||
| BC020771.1 | - | 4 | 561 | |||
| BC054002.1 | - | 4 | 530 | 6066745 | ||
| DB476861.2 | - | 4 | 644 | |||

FAM217B
Genomic Map