This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LLPH SIRNA Assays ›
Gene Symbol:
LLPH
Gene Name:
LLP homolog, long-term synaptic facilitation
Gene Aliases:
C12orf31, hLLP
Chromosome Location:
Chr.12: 66123069 - 66130753 on Build GRCh38
UniGene ID:
Hs.504820
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s227251

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 388.00
Your Price
Online offer: 287.12
287.12
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LLPH
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032338.3 NP_115714.1 3 285
GenBank mRNA AK057947.1 - 2 198
AK091568.1 - 3 285
BC006002.1 - 3 281 4247279
BC070232.1 - 3 277 4594274

Back To Top

More Information


Gene Ontology Categories:

Process(es)

poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-bn4dw:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline