This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CD70 SIRNA Assays ›
Gene Symbol:
CD70
Gene Name:
CD70 molecule
Gene Aliases:
CD27L, CD27LG, TNFSF7, TNLG8A
Chromosome Location:
Chr.19: 6581646 - 6591152 on Build GRCh38
UniGene ID:
Hs.501497
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s228530

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CD70
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252.4 NP_001243.1 3 587
XM_017027523.1 XP_016883012.1 3 549
GenBank mRNA AK298467.1 - 3 549
BC000725.2 - 3 533 3506629
BT007211.1 - 3 437
L08096.1 - 3 587
S69339.1 - 3 574

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

immune response
signal transduction
cell-cell signaling
cell proliferation
tumor necrosis factor-mediated signaling pathway
extrinsic apoptotic signaling pathway
protease binding
receptor binding
cytokine activity
tumor necrosis factor receptor binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-sjngx:80/100.66.77.211:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline