siRNA Details
| Gene Symbol: WDTC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276252.1 | NP_001263181.1 | 7 | 1181 | ||
| NM_015023.4 | NP_055838.2 | 7 | 1181 | |||
| XM_011541057.1 | XP_011539359.1 | 7 | 859 | |||
| XM_011541058.2 | XP_011539360.1 | 7 | 1126 | |||
| XM_017000730.1 | XP_016856219.1 | 7 | 1126 | |||
| GenBank mRNA | AB028960.1 | - | 2 | 320 | ||
| AK001734.1 | - | 6 | 653 | |||
| AK315564.1 | - | 7 | 1065 | |||
| BC032523.1 | - | 7 | 1044 | 5500384 | ||
| DQ572213.1 | - | 1 | 13 | |||

WDTC1
Genomic Map