siRNA Details
| Gene Symbol: C21orf91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100420.1 | NP_001093890.1 | 3 | 611 | ||
| NM_001100421.1 | NP_001093891.1 | 3 | 611 | |||
| NM_017447.3 | NP_059143.3 | 3 | 611 | |||
| GenBank mRNA | AF239726.1 | - | 3 | 564 | ||
| AK023825.1 | - | 3 | 576 | |||
| AK225123.1 | - | 3 | 582 | |||
| AK314471.1 | - | 3 | 584 | |||
| BC015468.2 | - | 2 | 527 | 3916462 | ||
| DB066982.1 | - | 1 | 326 | |||

C21orf91
Genomic Map