This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SCGB1C2 SIRNA Assays ›
Gene Symbol:
SCGB1C2 SCGB1C1
Gene Name:
secretoglobin family 1C member 2
secretoglobin family 1C member 1
Gene Aliases:
-
RYD5
Chromosome Location:
Chr.17: 132346 - 139067 on Build GRCh38
Chromosome Location:
Chr.11: 187881 - 194575 on Build GRCh38
UniGene ID:
Hs.127059
Hs.127059
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID s229380

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SCGB1C2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001097610.2 NP_001091079.1 3 298
XM_011535362.2 XP_011533664.1 4 578
Gene Symbol: SCGB1C1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145651.2 NP_663626.2 3 298
XM_005252804.3 XP_005252861.2 4 578
GenBank mRNA AY026938.1 - 3 298
BC069287.1 - 3 298 7262728
BC111427.1 - 3 297 40069184
BC111433.1 - 3 297 40069182

Back To Top

More Information


Panther Classification:

Molecular Function -

cytokine

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-xp8mr:80/100.66.76.90:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0