siRNA Details
| Gene Symbol: FNIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001323916.1 | NP_001310845.1 | 2 | 363 | ||
| NM_020840.2 | NP_065891.1 | 2 | 358 | |||
| XM_005263156.1 | XP_005263213.1 | 2 | 358 | |||
| XM_005263158.1 | XP_005263215.1 | 2 | 158 | |||
| XM_005263160.3 | XP_005263217.1 | 2 | 358 | |||
| XM_011532154.2 | XP_011530456.1 | 2 | 1138 | |||
| XM_017008487.1 | XP_016863976.1 | 2 | 358 | |||
| GenBank mRNA | AB040883.1 | - | 2 | 215 | ||
| AL713802.1 | - | 2 | 159 | |||
| BC016638.1 | - | 2 | 363 | |||
| DB482145.1 | - | 2 | 324 | |||
| HY049314.1 | - | 2 | 360 | |||

FNIP2
Genomic Map