This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RRP1B SIRNA Assays ›
Gene Symbol:
RRP1B
Gene Name:
ribosomal RNA processing 1B
Gene Aliases:
KIAA0179, NNP1L, Nnp1, PPP1R136, RRP1
Chromosome Location:
Chr.21: 43659551 - 43696079 on Build GRCh38
UniGene ID:
Hs.565725
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s22977

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RRP1B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015056.2 NP_055871.1 4, 5 465
GenBank mRNA AK056263.1 - 4, 5 428
BC014005.2 - 4, 5 437
BC028386.2 - 4, 5 470 4821672
D80001.1 - 4, 5 363

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

rRNA processing
negative regulation of phosphatase activity
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-smrv7:80/100.66.72.156:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0