siRNA Details
| Gene Symbol: Gm6121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114754.1 | NP_001108226.1 | 9 | 753 | ||
| Gene Symbol: Gm14525 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162364.2 | NP_001155836.1 | 9 | 753 | ||
| GenBank mRNA | BC048742.1 | - | 9 | 753 | ||

Gm6121
Genomic Map