This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OLFR1118 SIRNA Assays ›
Gene Symbol:
Olfr1118 Olfr1116
Gene Name:
olfactory receptor 1118
olfactory receptor 1116
Gene Aliases:
MOR264-22, Olfr1118-ps
MOR264-21P, MOR264-8P, Olfr1116-ps, Olfr1521-ps1
Chromosome Location:
Chr.2: 87308764 - 87309759 on Build GRCm38
Chromosome Location:
Chr.2: 87268846 - 87269769 on Build GRCm38
UniGene ID:
Mm.377942
Mm.424863
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s232029

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olfr1118
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207632.2 NP_997515.2 1 133
Gene Symbol: Olfr1116
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011734.1 NP_001011734.1 1 106

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception of smell
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-fjpjw:80/100.66.76.244:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0