siRNA Details
| Gene Symbol: Magi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170745.1 | NP_001164216.1 | 9 | 1956 | ||
| NM_001170746.1 | NP_001164217.1 | 10 | 2121 | |||
| NM_015823.3 | NP_056638.1 | 9 | 1956 | |||
| GenBank mRNA | AK039336.1 | - | 6 | 941 | ||
| AK122349.1 | - | 8 | 1416 | |||
| AK147530.1 | - | 9 | 1957 | |||
| BC059005.1 | - | 9 | 1613 | 6847238 | ||

Magi2
Genomic Map