This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GM14920 SIRNA Assays ›
Gene Symbol:
Gm14920 H2afb3
Gene Name:
predicted pseudogene 14920
H2A histone family, member B3
Gene Aliases:
ENSMUSG00000067441, Gm1539, H2A.Bbd1, OTTMUSG00000018481
EG624957, H2A.Bbd3, H2afb3-ps
Chromosome Location:
Chr.X: 117014757 - 117015104 on Build GRCm38
Chromosome Location:
Chr.X: 120312748 - 120313095 on Build GRCm38
UniGene ID:
Mm.469831
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s232169

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm14920
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102665.1 NP_001096135.1 1 253
Gene Symbol: H2afb3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281531.1 NP_001268460.1 1 253

Back To Top

More Information


Panther Classification:

Molecular Function -

histone

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-hssmq:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline