This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GM10058 SIRNA Assays ›
Gene Symbol:
Gm10058 Gm10096 Gm10147 Gm10230 Gm10486 Gm10487 Gm10488 Gm14525 Gm14632 Gm14819 Gm2012 Gm4836 Gm5169 Gm6121
Gene Name:
predicted gene 10058
predicted gene 10096
predicted gene 10147
predicted gene 10230
predicted gene 10486
predicted gene 10487
predicted gene 10488
predicted gene 14525
predicted gene 14632
predicted gene 14819
predicted gene 2012
predicted gene 4836
predicted gene 5169
predicted gene 6121
Gene Aliases:
ENSMUSG00000059047
ENSMUSG00000061227, Xmr-like
ENSMUSG00000064334
ENSMUSG00000068103
ENSMUSG00000073247
ENSMUSG00000073253
ENSMUSG00000073257
OTTMUSG00000017011
OTTMUSG00000017531
OTTMUSG00000018086
-
-
Xmr
-
Chromosome Location:
Chr.X: 27924021 - 27946885 on Build GRCm38
Chromosome Location:
Chr.X: 28931807 - 28954702 on Build GRCm38
Chromosome Location:
Chr.X: 28584995 - 28607874 on Build GRCm38
Chromosome Location:
Chr.X: 27472033 - 27494914 on Build GRCm38
Chromosome Location:
Chr.X: 35067384 - 35090316 on Build GRCm38
Chromosome Location:
Chr.X: 30820542 - 30843469 on Build GRCm38
Chromosome Location:
Chr.X: 29976314 - 29999162 on Build GRCm38
Chromosome Location:
Chr.X: 26672780 - 26702638 on Build GRCm38
Chromosome Location:
Chr.X: 30329715 - 30352646 on Build GRCm38
Chromosome Location:
Chr.X: 35404215 - 35427094 on Build GRCm38
Chromosome Location:
Chr.X: 26200882 - 26231323 on Build GRCm38
Chromosome Location:
Chr.X: 28270710 - 28293632 on Build GRCm38
Chromosome Location:
Chr.X: 25277487 - 25301455 on Build GRCm38
Chromosome Location:
Chr.X: 26912743 - 26935669 on Build GRCm38
UniGene ID:
Mm.483167
-
-
Mm.483167
Mm.458011
Mm.427330
Mm.432195
Mm.379326
Mm.483167
Mm.483167
Mm.484125
Mm.427330
Mm.379326
Mm.483167
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
-
-
-
-
-
-
-
-
-
-
-
-
siRNA ID s232223

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm10058
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109969.2 NP_001103439.1 2 191
GenBank mRNA BU936093.1 - 194
Gene Symbol: Gm10096
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102678.2 NP_001096148.1 2 222
Gene Symbol: Gm10147
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099919.2 NP_001093389.1 194
Gene Symbol: Gm10230
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099347.2 NP_001092817.1 2 191
Gene Symbol: Gm10486
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109970.1 NP_001103440.1 2 222
GenBank mRNA BC087930.1 - 191 6773503
BC099540.1 - 2 162 6774523
X72697.1 - 209
Gene Symbol: Gm10487
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100609.1 NP_001094079.1 2 222
Gene Symbol: Gm10488
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099325.2 NP_001092795.1 2 191
GenBank mRNA BC099541.1 - 2 191 30916030
Gene Symbol: Gm14525
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162364.2 NP_001155836.1 2 191
GenBank mRNA BC048742.1 - 2 191
Gene Symbol: Gm14632
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100610.2 NP_001094080.1 2 191
Gene Symbol: Gm14819
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110250.2 NP_001103720.1 191
Gene Symbol: Gm2012
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104946.2 NP_001098416.1 2 191
Gene Symbol: Gm4836
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009529.3 NP_033555.1 2 222
GenBank mRNA BC061169.1 - 2 222 6774630
Gene Symbol: Gm5169
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040669.1 NP_001035759.1 2 188
GenBank mRNA BC087929.1 - 2 188 6742946
Gene Symbol: Gm6121
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114754.1 NP_001108226.1 2 191

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

spermatid development
biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline