This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OSBPL7 SIRNA Assays ›
Gene Symbol:
Osbpl7
Gene Name:
oxysterol binding protein-like 7
Gene Aliases:
4933437E18Rik
Chromosome Location:
Chr.11: 97050344 - 97068904 on Build GRCm38
UniGene ID:
Mm.190450
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s232231

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Osbpl7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081434.1 NP_001074903.1 15 1611
GenBank mRNA AK154677.1 - 15 1647
AK171585.1 - 15 1640

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cellular response to cholesterol
positive regulation of proteasomal protein catabolic process
cholesterol binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0