This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZFP935 SIRNA Assays ›
Gene Symbol:
Zfp935
Gene Name:
zinc finger protein 935
Gene Aliases:
8430426H19Rik, Es113
Chromosome Location:
Chr.13: 62453016 - 62466812 on Build GRCm38
UniGene ID:
Mm.442497
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s232587

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp935
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136496.1 NP_001129968.1 3 975
NM_178875.4 NP_849206.2 4 1036
GenBank mRNA AK018441.1 - 3 966
AY189975.1 - 3 952
BC037448.1 - 3 980
BC047396.1 - 4 1013 3979342
BC087936.1 - 4 1036
BC113193.1 - 4 1032 40062446
BC115966.1 - 894

Back To Top

More Information


Panther Classification:

Molecular Function -

DNA-binding transcription factor zinc finger transcription factor C2H2 zinc finger transcription factor gene-specific transcriptional regulator

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline