This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GM10488 SIRNA Assays ›
Gene Symbol:
Gm10488 Gm10058 Gm10096 Gm10147 Gm10230 Gm10486 Gm10487 Gm14632 Gm14819 Gm4836
Gene Name:
predicted gene 10488
predicted gene 10058
predicted gene 10096
predicted gene 10147
predicted gene 10230
predicted gene 10486
predicted gene 10487
predicted gene 14632
predicted gene 14819
predicted gene 4836
Gene Aliases:
ENSMUSG00000073257
ENSMUSG00000059047
ENSMUSG00000061227, Xmr-like
ENSMUSG00000064334
ENSMUSG00000068103
ENSMUSG00000073247
ENSMUSG00000073253
OTTMUSG00000017531
OTTMUSG00000018086
-
Chromosome Location:
Chr.X: 29976314 - 29999162 on Build GRCm38
Chromosome Location:
Chr.X: 27924021 - 27946885 on Build GRCm38
Chromosome Location:
Chr.X: 28931807 - 28954702 on Build GRCm38
Chromosome Location:
Chr.X: 28584995 - 28607874 on Build GRCm38
Chromosome Location:
Chr.X: 27472033 - 27494914 on Build GRCm38
Chromosome Location:
Chr.X: 35067384 - 35090316 on Build GRCm38
Chromosome Location:
Chr.X: 30820542 - 30843469 on Build GRCm38
Chromosome Location:
Chr.X: 30329715 - 30352646 on Build GRCm38
Chromosome Location:
Chr.X: 35404215 - 35427094 on Build GRCm38
Chromosome Location:
Chr.X: 28270710 - 28293632 on Build GRCm38
UniGene ID:
Mm.432195
Mm.483167
-
-
Mm.483167
Mm.458011
Mm.427330
Mm.483167
Mm.483167
Mm.427330
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
-
-
-
-
-
-
-
-
siRNA ID s232742

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 388.65
388.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm10488
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099325.2 NP_001092795.1 8 663
GenBank mRNA BC099541.1 - 8 663 30916030
Gene Symbol: Gm10058
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109969.2 NP_001103439.1 8 663
GenBank mRNA BU936093.1 - 666
Gene Symbol: Gm10096
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102678.2 NP_001096148.1 8 694
Gene Symbol: Gm10147
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099919.2 NP_001093389.1 666
Gene Symbol: Gm10230
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099347.2 NP_001092817.1 8 663
Gene Symbol: Gm10486
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109970.1 NP_001103440.1 8 694
GenBank mRNA BC087930.1 - 663 6773503
BC099540.1 - 8 634 6774523
X72697.1 - 681
Gene Symbol: Gm10487
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100609.1 NP_001094079.1 8 694
Gene Symbol: Gm14632
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100610.2 NP_001094080.1 8 663
Gene Symbol: Gm14819
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110250.2 NP_001103720.1 663
Gene Symbol: Gm4836
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009529.3 NP_033555.1 8 694
GenBank mRNA BC061169.1 - 8 694 6774630

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

spermatid development
biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-474fd:80/100.66.77.40:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0