siRNA Details
| Gene Symbol: Znhit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027318.3 | NP_081594.1 | 5 | 919 | ||
| GenBank mRNA | AK012199.1 | - | 5 | 559 | ||
| AK080799.1 | - | 5 | 920 | |||
| BC026751.1 | - | 5 | 565 | 3153902 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Znhit1| Gene Symbol: Znhit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027318.3 | NP_081594.1 | 5 | 919 | ||
| GenBank mRNA | AK012199.1 | - | 5 | 559 | ||
| AK080799.1 | - | 5 | 920 | |||
| BC026751.1 | - | 5 | 565 | 3153902 | ||