siRNA Details
| Gene Symbol: Scgb1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099742.1 | NP_001093212.1 | 2 | 85 | ||
| GenBank mRNA | AA462242.1 | - | 2 | 61 | ||
| BY000447.1 | - | 2 | 86 | |||
| CB173308.1 | - | 2 | 62 | |||
| CB174369.1 | - | 2 | 57 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scgb1c1| Gene Symbol: Scgb1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099742.1 | NP_001093212.1 | 2 | 85 | ||
| GenBank mRNA | AA462242.1 | - | 2 | 61 | ||
| BY000447.1 | - | 2 | 86 | |||
| CB173308.1 | - | 2 | 62 | |||
| CB174369.1 | - | 2 | 57 | |||