siRNA Details
| Gene Symbol: Zfp874a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177712.4 | NP_808380.1 | 4 | 633 | ||
| GenBank mRNA | AK049183.1 | - | 4 | 621 | ||
| AK145787.1 | - | 4 | 633 | |||
| BC080824.1 | - | 712 | 30458333 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp874a| Gene Symbol: Zfp874a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177712.4 | NP_808380.1 | 4 | 633 | ||
| GenBank mRNA | AK049183.1 | - | 4 | 621 | ||
| AK145787.1 | - | 4 | 633 | |||
| BC080824.1 | - | 712 | 30458333 | |||