This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ASTN2 SIRNA Assays ›
Gene Symbol:
ASTN2
Gene Name:
astrotactin 2
Gene Aliases:
bA67K19.1
Chromosome Location:
Chr.9: 116425225 - 117415039 on Build GRCh38
UniGene ID:
Hs.601562
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s23382

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ASTN2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184734.1 NP_001171663.1 7 1114
NM_014010.4 NP_054729.3 21 3670
NM_198186.3 NP_937829.3 7 1435
NM_198187.3 NP_937830.3 7 1114
NM_198188.2 NP_937831.1 7 1114
GenBank mRNA AB014534.1 - 21 3670
AF116574.1 - 19 2891
AK024064.1 - 7 1085
AK127350.1 - 20 2891
AK315025.1 - 19 2734
BC018759.2 - 15 2119
BC029272.1 - 7 1435
BC093835.1 - 7 948
BC101667.1 - 7 947
BC143289.1 - 7 948
BC143292.1 - 7 948
BC143293.1 - 7 948
BC146756.1 - 21 3670 9007192
BX649176.1 - 21 3005

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001271363.
This siRNA also targets the following transcripts in Mouse: NM_207109, NM_019514, AK041160, AF116573.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein localization to cell surface
calcium ion binding
inositol 1,3,4,5 tetrakisphosphate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline