This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other H3F3A SIRNA Assays ›
Gene Symbol:
H3f3a
Gene Name:
H3 histone, family 3A
Gene Aliases:
H3.3A
Chromosome Location:
Chr.1: 180802568 - 180813603 on Build GRCm38
UniGene ID:
Mm.138832
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s234173

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: H3f3a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008210.4 NP_032236.1 4 696
GenBank mRNA AK008405.1 - 3 597
AK037900.1 - 4 698
AK150467.1 - 4 656
AK150468.1 - 4 656
AK150591.1 - 4 651
AK150928.1 - 4 651
AK151634.1 - 4 656
AK152293.1 - 4 651
AK153034.1 - 4 655
AK160677.1 - 4 655
AK162040.1 - 4 654
AK167879.1 - 4 655
AK168197.1 - 4 655
AK169608.1 - 4 651
AK187043.1 - 1 190
BC002268.1 - 4 641 3495830
BC012687.1 - 4 645 4224280
BC083353.1 - 4 636 5714859
BC088835.1 - 4 633 6437687
BC106177.1 - 4 630 6511787
Z85979.1 - 4 644
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BC027275.1 - 2171 Chr. - - [-] on Build

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_003750348, XM_002729644, XM_002728043, FQ218544, FQ220271, and others.

Panther Classification:

Molecular Function -

histone

Gene Ontology Categories:

Function(s) Process(es)

osteoblast differentiation
muscle cell differentiation
negative regulation of chromosome condensation
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
nucleosomal DNA binding
histone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-smrv7:80/100.66.72.156:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0