siRNA Details
| Gene Symbol: Gm11569 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099312.1 | NP_001092782.1 | 1 | 757 | ||
| Gene Symbol: Krtap4-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027087.3 | NP_081363.1 | 1 | 716 | ||
| GenBank mRNA | AK009035.1 | - | 1 | 717 | ||

Gm11569
Genomic Map