This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GM11569 SIRNA Assays ›
Gene Symbol:
Gm11569 Krtap4-13
Gene Name:
predicted gene 11569
keratin associated protein 4-13
Gene Aliases:
OTTMUSG00000002208
2300006N05Rik
Chromosome Location:
Chr.11: 99798064 - 99798932 on Build GRCm38
Chromosome Location:
Chr.11: 99809078 - 99809896 on Build GRCm38
UniGene ID:
Mm.425705
Mm.158279
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s234608

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm11569
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099312.1 NP_001092782.1 1 789
Gene Symbol: Krtap4-13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027087.3 NP_081363.1 1 748
GenBank mRNA AK009035.1 - 1 749

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-fbxsq:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline