This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DMPK SIRNA Assays ›
Gene Symbol:
Dmpk
Gene Name:
dystrophia myotonica-protein kinase
Gene Aliases:
Dm15
Chromosome Location:
Chr.1: 79989020 - 79999358 on Build Rnor_6.0
UniGene ID:
Rn.130535
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s234692

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dmpk
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759040.1 XP_008757262.1 14 2202
XM_008759041.1 XP_008757263.1 14 2206
XM_008759042.1 XP_008757264.1 14 2187
XM_008759043.1 XP_008757265.1 15 2036
XM_008759044.1 XP_008757266.1 15 2040
XM_008759046.1 XP_008757268.1 15 2021
XM_008759047.1 XP_008757269.1 15 2025
XM_008759048.1 XP_008757270.1 14 3278
XM_008759051.1 XP_008757273.1 16 2101
GenBank mRNA FQ217505.1 - 14 1837
FQ224460.1 - 14 1852

Back To Top

More Information


Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

regulation of sodium ion transport
protein phosphorylation
cellular calcium ion homeostasis
nuclear envelope organization
regulation of heart contraction
biological_process
muscle cell apoptotic process
regulation of myotube differentiation
regulation of skeletal muscle contraction by calcium ion signaling
regulation of excitatory postsynaptic membrane potential involved in skeletal muscle contraction
regulation of catalytic activity
regulation of synapse structural plasticity
molecular_function
protein serine/threonine kinase activity
ATP binding
myosin phosphatase regulator activity
heat shock protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-xql92:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline