siRNA Details
| Gene Symbol: LOC688924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109519.1 | NP_001102989.1 | 3 | 153 | ||
| XM_008759189.1 | XP_008757411.1 | 3 | 195 | |||
| XM_008759190.1 | XP_008757412.1 | 3 | 183 | |||
| XM_008759191.1 | XP_008757413.1 | 4 | 223 | |||
| XM_008759192.1 | XP_008757414.1 | 4 | 339 | |||
| XM_008759193.1 | XP_008757415.1 | 3 | 166 | |||
| XM_008759194.1 | XP_008757416.1 | 3 | 258 | |||
| XM_008759195.1 | XP_008757417.1 | 3 | 142 | |||

LOC688924
Genomic Map